Learn More
Ki-67, Biotin, clone: 20Raj1, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 13-5699-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Ki-67 is a nuclear protein that is expressed during various stages in the cell cycle, particularly during late G1, S, G2, and M phases. The protein has a forkhead associated domain (FHA) through which it associates with the euchromatin at the perichromosomal layer with centromeric heterochromatin, and with the nucleolus. Ki-67 is shown to have a cell cycle dependent topographical distribution, with perinucleolar expression at G1, expression in the nuclear matrix at G2, and expression on the chromosomes during M phase. Ki-67 is commonly used as a proliferation marker because it is not detected in G0 cells, but increases steadily from G1 through mitosis.
Description: The monoclonal antibody 20Raj1 recognizes the human Ki-67 protein. Two isoforms of Ki-67 exist, a 345 and 395kDa form that are expressed in dividing cells. Ki-67 is expressed in all cell types and is detectable during active phases of the cell cycle (G1, S, G2, and mitosis) but is absent from resting cells (G0). During interphase, Ki-67 expression is localized to the nucleus but redistributes to the chromosomes during mitosis and has specifically been found to associate with heterochromatin-bound proteins such as chromobox protein homolog 3 (CBX3). In studies of tumor cells, Ki-67 expression has been used as a marker for determining the fraction of proliferating cells within a given population of tumor cells.This monoclonal antibody 20Raj1 recognizes canine Ki-67.Applications Reported: This 20Raj1 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.Applications Tested: This 20Raj1 antibody has been tested by intracellular staining of normal human peripheral blood cells using the Foxp3/Transcription Factor Buffers Set (cat. 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The 20Raj1 antibody has also been tested by immunocytochemistry of fixed and permeablilized cells and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.Specifications
Ki-67 | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P46013 | |
MKI67 | |
Affinity chromatography | |
RUO | |
100686578, 4288 | |
Store at 2-8°C. Do not freeze. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
20Raj1 | |
Biotin | |
MKI67 | |
antigen identified by monoclonal antibody Ki-67, antigen KI-67, KIA, proliferation-related Ki-67 antigen, protein phosphatase 1, regulatory subunit 105, RP11-380J17.2 | |
Mouse | |
25 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 κ |
For Research Use Only.