missing translation for 'onlineSavingsMsg'
Learn More

KH 7, Tocris Bioscience™

Selective soluble adenylyl cyclase inhibitor

Brand:  Tocris (Bio-Techne) 383410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463674

  • £155.76 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Selective soluble adenylyl cyclase (sAC) inhibitor (IC50 = 3 - 10μM in vivo). Inert towards transmembrane adenylyl cyclase (tmAC) in vitro and in whole cells at concentrations up to 300μM. Blocks synthesis of cAMP and displays an antiapoptotic effect at concentrations of 1 - 100μM.
Specifications

Chemical Identifiers

330676-02-3
419.297
kh 7
(2S)-2-(1H-benzimidazol-2-ylsulfanyl)-N'-[(Z)-(3-bromo-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]propanehydrazide
C17H15BrN4O2S
GEHVZUUHMWNMAY-RGAIUGGKSA-N
98069644
CC(C(=O)NNC=C1C=C(C=CC1=O)Br)SC2=NC3=CC=CC=C3N2

Specifications

KH 7
330676-02-3
kh 7
CC(C(=O)NNC=C1C=C(C=CC1=O)Br)SC2=NC3=CC=CC=C3N2
419.297
419.3
10mg
C17H15BrN4O2S
GEHVZUUHMWNMAY-RGAIUGGKSA-N
(2S)-2-(1H-benzimidazol-2-ylsulfanyl)-N'-[(Z)-(3-bromo-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]propanehydrazide
98069644
>99%
Videos