missing translation for 'onlineSavingsMsg'
Learn More

Ketanserin tartrate, Tocris Bioscience™

Selective 5-HT2A antagonist. Also antagonist at 5-HT1D

Brand:  Tocris (Bio-Techne) 0908/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458143

  • £114.75 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Selective 5-HT2A serotonin receptor antagonist; can also be used to discriminate between 5-HT1D and 5-HT1B receptor subtypes.
Specifications

Chemical Identifiers

83846-83-7
545.52
3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2, 4[1H, 3H]-quinazolinedione tartrate
C26H28FN3O9
KMTLTEVOQLMYRS-WUUYCOTASA-N
91885469

Specifications

Ketanserin tartrate
>97%
3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2, 4[1H, 3H]-quinazolinedione tartrate
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O.C(C(C(=O)O)O)(C(=O)O)O
545.52
545.52
83846-83-7
C26H28FN3O9
KMTLTEVOQLMYRS-WUUYCOTASA-N
(2S,3S)-2,3-dihydroxybutanedioic acid;3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
91885469
50mg
Videos