missing translation for 'onlineSavingsMsg'
Learn More

Ketanserin tartrate, Tocris Bioscience™

Selective 5-HT2A antagonist. Also antagonist at 5-HT1D

Brand:  Tocris (Bio-Techne) 0908/250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 250mg

 View more versions of this product

Product Code. 13448143

  • £439.53 / 250mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective 5-HT2A serotonin receptor antagonist; can also be used to discriminate between 5-HT1D and 5-HT1B receptor subtypes.
Specifications

Chemical Identifiers

83846-83-7
545.52
ketanserin tartrate
(2S,3S)-2,3-dihydroxybutanedioic acid;3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
C26H28FN3O9
KMTLTEVOQLMYRS-WUUYCOTASA-N
91885469
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O.C(C(C(=O)O)O)(C(=O)O)O

Specifications

250mg
83846-83-7
ketanserin tartrate
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O.C(C(C(=O)O)O)(C(=O)O)O
545.52
545.52
Ketanserin tartrate
C26H28FN3O9
KMTLTEVOQLMYRS-WUUYCOTASA-N
(2S,3S)-2,3-dihydroxybutanedioic acid;3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
91885469
>97%
Videos
Documents