missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kerosene, purum p.a, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Glass bottle
Specifications
Color | Colorless |
Density | 0.8g/mL at 25°C |
Boiling Point | 175° to 3257deg;C |
Packaging | Glass bottle |
Quantity | 1 L |
UN Number | UN1223 |
Vapor Density | 4.5 (vs air) |
Physical Form | Liquid |
Chemical Name or Material | Kerosene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'