Learn More
Duran™ Keck Hose Clamps
Control the amount of a substance flowing through the hose using only one finger
Brand: Duran™ 286338307
Code : A0
Additional Details : Weight : 0.00900kg
Description
- Amount of a substance flowing through the hose can be controlled precisely using only one finger
- Thermoplastic polyester (PBT) clamps are temperature resistant up to 200°C
- Autoclavable