Learn More
KDM6A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551521
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SNSCILLDKC PPPRPPTSPY PPLPKDKLNP PTPSIYLENK RDAFFPPLHQ FCTNPKNPVT VIRGLAGALK LDLGLFSTKT LVEANNEHMV EVRTQLLQPA DENWDPTGTK KIWRCESNRS HTTI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
KDM6A Polyclonal antibody specifically detects KDM6A in Human samples. It is validated for Immunohistochemistry.Specifications
KDM6A | |
Polyclonal | |
Unconjugated | |
KDM6A | |
4lysine (K)-specific demethylase 6A; bA386N14.2; bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)); histone demethylase UTX; KABUK2; Kdm6a; lysine (K)-specific demethylase 6A; lysine demethylase 6A; lysine-specific demethylase 6A; lysine-specific demethylase 6A-like protein; NEWGENE_1565481; ubiqiutous TPR motif protein; ubiquitous TPR motif protein UTX; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat protein X-linked; ubiquitously transcribed tetratricopeptide repeat, X chromosome; ubiquitously transcribed TPR protein on the X chromosome; Ubiquitously transcribed X chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR gene on the X chromosome; Ubiquitously-transcribed TPR protein on the X chromosome; Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein; UTX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7403 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15550 | |
KDM6A | |
Recombinant protein corresponding to Human KDM6A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only