missing translation for 'onlineSavingsMsg'
Learn More

KD Scientific Stainless-Steel Syringes

Durable, electron-beam construction withstands high force, high pressure applications

Brand:  KD Scientific 780804

Code : A0

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 11986706

  • £875.16 / Each


Estimated Shipment
07-06-2018
Add to basket

Description

Description

Eliminates hazards of glass syringe breakage.

  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals

Specifications

Specifications

31.7mm
50mL
Swagelok 1/16 in. Syringe
1500psi
28.4mm
316 Stainless Steel
Syringe pump
Videos