missing translation for 'onlineSavingsMsg'
Learn More

KD Scientific Dual and Multiple Syringe Infusion/Withdrawal Pumps

Easy-to-use programmable syringe pumps

Brand:  KD Scientific 789210


Additional Details : Weight : 4.00000kg

 View more versions of this product

Product Code. 10168514

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Simple menu-driven setup to program syringe diameter, flow rate and dispense volume
  • Backlit display
  • Automatic shutoff
  • Settings can be reviewed and changed during operation
  • Last settings are stored in memory for repeatability
Multiple Syringe Pumps
  • Capacity: 10 μL to 140 mL up to 10 syringes (depending on syringe size)
  • Built-in RS-232-C and TTL interfaces for easy external control
  • Hold glass or plastic syringes
  • Available as infusion and infusion/withdrawal models
Specifications

Specifications

115 V
10 μL to 140 mL
18 kg
±0.2%
Backlit LCD
2
0.001 μL/hr. to 147 mL/min.
RS232C, TTL
40 lb.
Infusion Withdrawal Pump
±0.5%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers