Learn More
KCTD16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562146
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PQETVICGPV TRQTNIQTLD RPIKKGPVQL IQQSEMRRKS DLLRTLTSGS RESNMSSKKK AVKEKLS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 27%.
The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KCTD16 (potassium channel tetramerisation domain containing 16), also known as BTB/POZ domain-containing protein KCTD16, is a 428 amino acid protein that contains one BTB (POZ) domain. An auxiliary subunit of GABAB R1 and GABAB R2, KCTD16 increases agonist potency and alters the G-protein signaling of the receptors by accelerating onset and promoting desensitization.Specifications
KCTD16 | |
Polyclonal | |
Unconjugated | |
KCTD16 | |
4930434H12Rik; BTB/POZ domain-containing protein KCTD16; Gm1267; KCTD16; KIAA1317; potassium channel tetramerisation domain containing 16; potassium channel tetramerization domain containing 16; potassium channel tetramerization domain-containing protein 16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57528 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68DU8 | |
KCTD16 | |
Recombinant protein corresponding to Human KCTD16 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only