Learn More
KCNE1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559946
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MNCSESQRLR TLLSRLLLEL HHRGNASGLG AGPRPSMGMG VVPDPFVGRE VTSAKGDDA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
KCNE1L Polyclonal antibody specifically detects KCNE1L in Human samples. It is validated for Immunohistochemistry.Specifications
KCNE1L | |
Polyclonal | |
Unconjugated | |
KCNE5 | |
1500015C14Rik; AMME syndrome candidate gene 2 protein; AMMECR2; AMMECR2 protein; cardiac voltage-gated potassium channel accessory subunit 5; KCNE1L; KCNE1-like; Kcne5; Mink; MinK-like protein; potassium channel subunit beta MiRP4; potassium channel, voltage gated subfamily E regulatory beta subunit 5; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 5; potassium voltage-gated channel subfamily E member 1-like protein; Potassium voltage-gated channel subfamily E regulatory beta subunit 5; potassium voltage-gated channel subfamily E regulatory subunit 5; potassium voltage-gated channel, Isk-related family, member 1-like; potassium voltage-gated channel, Isk-related family, member 1-like, pseudogene; voltage-gated potassium channel accessory subunit 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23630 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJ90 | |
KCNE5 | |
Recombinant protein corresponding to Human KCNE1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only