missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KBTBD6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561836
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TGPEELKDTA HSAALLAQLK SFYDARLLCD VTIEVVTPGS GPGTGRLFPC NRNVLAAACP YFKSM Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
KBTBD6 Polyclonal antibody specifically detects KBTBD6 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
KBTBD6 | |
Polyclonal | |
Unconjugated | |
KBTBD6 | |
KBTBD6; kelch repeat and BTB (POZ) domain containing 6; kelch repeat and BTB domain containing 6; kelch repeat and BTB domain-containing protein 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89890 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86V97 | |
KBTBD6 | |
Recombinant protein corresponding to Human KBTBD6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only