missing translation for 'onlineSavingsMsg'
Learn More

kb NB 142-70, Tocris Bioscience™

Selective PKD inhibitor; analog of CID 755673 (Cat. No. 3327)

Brand:  Tocris (Bio-Techne) 396250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13443894

  • £829.40 / 50mg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective protein kinase D (PKD) inhibitor (IC50 values are 28.3, 58.7 and 53.2 nM for PKD1, 2 and 3 respectively). More potent analog of CID 755673 (Cat. No. 3327) with 7-fold greater inhibition. Inhibits prostrate cancer cell migration and invasion and reduces wound healing in vitro; displays prominent cytotoxic and anti-proliferative effects.
Specifications

Chemical Identifiers

1233533-04-4
251.318
3,4-dihydro-9-hydroxy-1 benzothieno 2,3-f-1,4-thiazepin-5 2h-one, 9-hydroxy-3,4-dihydro-2h-1 benzothiolo 2,3-f 1,4 thiazepin-5-one, 3,4-dihydro-9-hydroxy-1 benzothieno-2,3-f-1,4-thiazepin-5 2h-one, 4-hydroxy-8,14-dithia-11-azatricyclo 7.5.0.0 2 ,? tetradeca-1 9 ,2,4,6-tetraen-10-one, qcr-15, kb-nb hplc, 9-hydroxy-3,4-dihydro-2h-1-benzothiolo 2,3-f 1,4 thiazepin-5-one, 9-hydroxy-3,4-dihydro 1 benzothieno 2,3-f 1,4 thiazepin-5 2h-one
9-hydroxy-3,4-dihydro-2H-[1]benzothiolo[2,3-f][1,4]thiazepin-5-one
C11H9NO2S2
DHUAGGSHTKPOHU-UHFFFAOYSA-N
45258277
C1CSC2=C(C(=O)N1)SC3=C2C=C(C=C3)O

Specifications

kb NB 142-70
1233533-04-4
3,4-dihydro-9-hydroxy-1 benzothieno 2,3-f-1,4-thiazepin-5 2h-one, 9-hydroxy-3,4-dihydro-2h-1 benzothiolo 2,3-f 1,4 thiazepin-5-one, 3,4-dihydro-9-hydroxy-1 benzothieno-2,3-f-1,4-thiazepin-5 2h-one, 4-hydroxy-8,14-dithia-11-azatricyclo 7.5.0.0 2 ,? tetradeca-1 9 ,2,4,6-tetraen-10-one, qcr-15, kb-nb hplc, 9-hydroxy-3,4-dihydro-2h-1-benzothiolo 2,3-f 1,4 thiazepin-5-one, 9-hydroxy-3,4-dihydro 1 benzothieno 2,3-f 1,4 thiazepin-5 2h-one
C1CSC2=C(C(=O)N1)SC3=C2C=C(C=C3)O
251.318
251.32
50mg
C11H9NO2S2
DHUAGGSHTKPOHU-UHFFFAOYSA-N
9-hydroxy-3,4-dihydro-2H-[1]benzothiolo[2,3-f][1,4]thiazepin-5-one
45258277
>98%
Videos