missing translation for 'onlineSavingsMsg'
Learn More

kappa-Carrageenan, Thermo Scientific Chemicals

Catalog Number p-7053472
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
11114-20-8
C24H36O25S2-2
788.647
MFCD00151514
ZNOZWUKQPJXOIG-XSBHQQIPSA-L
kappa-carrageenan, kappa-carrageenans, 1->4-3,6-anhydro-alpha-d-galactopyranosyl-1->3-4-o-sulfonato-beta-d-galactopyranan
11966249
[(2R,3S,4R,5R,6S)-6-[[(1R,3S,4R,5R,8S)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-[[(1R,3R,4R,5R,8S)-8-[(2S,3R,4R,5R,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-sulfonatooxyoxan-2-yl]oxy-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-3-yl]oxy]-5-hydroxy-2-(
C1C2C(C(O1)C(C(O2)O)O)OC3C(C(C(C(O3)CO)OS(=O)(=O)[O-])OC4C(C5C(C(O4)CO5)OC6C(C(C(C(O6)CO)OS(=O)(=O)[O-])O)O)O)O
This item is not returnable. View return policy
11114-20-8
C24H36O25S2-2
788.647
MFCD00151514
ZNOZWUKQPJXOIG-XSBHQQIPSA-L
kappa-carrageenan, kappa-carrageenans, 1->4-3,6-anhydro-alpha-d-galactopyranosyl-1->3-4-o-sulfonato-beta-d-galactopyranan
11966249
[(2R,3S,4R,5R,6S)-6-[[(1R,3S,4R,5R,8S)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-[[(1R,3R,4R,5R,8S)-8-[(2S,3R,4R,5R,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-sulfonatooxyoxan-2-yl]oxy-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-3-yl]oxy]-5-hydroxy-2-(
C1C2C(C(O1)C(C(O2)O)O)OC3C(C(C(C(O3)CO)OS(=O)(=O)[O-])OC4C(C5C(C(O4)CO5)OC6C(C(C(C(O6)CO)OS(=O)(=O)[O-])O)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 5 g
Merck Index 14,1864
Formula Weight 401.32
Physical Form Solid
Chemical Name or Material kappa-Carrageenan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'