missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 14 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Kallikrein 14 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization, ELISA |
Classification | Polyclonal |
Description
Kallikrein 14 Polyclonal specifically detects Kallikrein 14 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
Kallikrein 14 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Polyclonal | |
Goat | |
Human | |
EC 3.4.21, EC 3.4.21.-, hK14, kallikrein 14, Kallikrein-like protein 6, kallikrein-related peptidase 14, KLK14, KLKL6, KLK-L6kallikrein-14 | |
KLK14 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation, Neutralization, ELISA | |
Unconjugated | |
RUO | |
AAD50773 | |
43847 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 14 Gln19-Met248, predicted Accession # AAD50773 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Kallikrein 14 in ELISAs and Western blots. |
For Research Use Only