missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 12 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Kallikrein 12 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Kallikrein 12 Ala18-Asn248 Accession # Q9UKR0 |
Gene ID (Entrez) | 43849 |
Gene Accession No. | Q9UKR0 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
Kallikrein 12 Polyclonal specifically detects Kallikrein 12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Kallikrein 12 | |
43849 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Kallikrein 12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Kallikrein 8, rhKallikrein 10, rhKallikrein 11, and rhKallikrein 15 is observed. | |
Unconjugated | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 12 Ala18-Asn248 Accession # Q9UKR0 | |
Q9UKR0 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Sheep | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only