Learn More
Kartell™ K70 Silicon Hoses
Brand:  Kartell™ 3928
Code : NEW
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10207301
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
FDA, USP, Class VI, A 50p60 and EPC VI.1.3.2 Approved
Silicone
10m
–60°C
9mm
13mm
200°C
Videos