missing translation for 'onlineSavingsMsg'
Learn More

K 579, Tocris Bioscience™

Dipeptidyl peptidase IV (DPP-IV) inhibitor

Brand:  Tocris (Bio-Techne) 279010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441844

  • £235.20 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Dipeptidyl peptidase IV inhibitor. Inhibits rat, canine, human and monkey enzymes with IC50 values of 3, 5, 8, and 8 nM respectively. Long-acting hypoglycemic agent; attenuates glucose excursion following glucose loading in Zucker fatty rats.
Specifications

Chemical Identifiers

440100-64-1
328.42
unii-74p4vv90ru, dipeptidylpeptidase iv inhibitor iv, k579, 2s-1-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl-2-pyrrolidinecarbonitrile, 2-pyrrolidinecarbonitrile, 1-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl, 2-pyrrolidinecarbonitrile, 1-2-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl-, 2s, s-1-4-methyl-1-pyrimidin-2-yl piperidin-4-yl glycyl pyrrolidine-2-carbonitrile
(2S)-1-[2-[(4-methyl-1-pyrimidin-2-ylpiperidin-4-yl)amino]acetyl]pyrrolidine-2-carbonitrile
C17H24N6O
JERPHRNGJBSFAP-AWEZNQCLSA-N
9818824
CC1(CCN(CC1)C2=NC=CC=N2)NCC(=O)N3CCCC3C#N

Specifications

10mg
440100-64-1
unii-74p4vv90ru, dipeptidylpeptidase iv inhibitor iv, k579, 2s-1-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl-2-pyrrolidinecarbonitrile, 2-pyrrolidinecarbonitrile, 1-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl, 2-pyrrolidinecarbonitrile, 1-2-4-methyl-1-2-pyrimidinyl-4-piperidinyl amino acetyl-, 2s, s-1-4-methyl-1-pyrimidin-2-yl piperidin-4-yl glycyl pyrrolidine-2-carbonitrile
CC1(CCN(CC1)C2=NC=CC=N2)NCC(=O)N3CCCC3C#N
328.42
328.41
K 579
C17H24N6O
JERPHRNGJBSFAP-AWEZNQCLSA-N
(2S)-1-[2-[(4-methyl-1-pyrimidin-2-ylpiperidin-4-yl)amino]acetyl]pyrrolidine-2-carbonitrile
9818824
>99%
Videos
Documents