missing translation for 'onlineSavingsMsg'
Learn More

JX 401, Tocris Bioscience™

Potent, reversible p38α inhibitor

Brand:  Tocris (Bio-Techne) 265710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13451664

  • £208.74 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, reversible inhibitor of p38α that displays no activity on the p38γ isoform (IC50 values are 32 nM and > 10 μM respectively). Inhibits the differentiation of myoblasts to myotubes in mammalian cells in culture.
Specifications

Chemical Identifiers

349087-34-9
355.496
unii-tbu7utv30q, tbu7utv30q, 4-benzylpiperidin-1-yl 2-methoxy-4-methylsulfanyl phenyl methanone, 4-benzyl-1-2-methoxy-4-methylsulfanyl benzoyl piperidine, piperidine, 1-2-methoxy-4-methylthio benzoyl-4-phenylmethyl, methanone, 2-methoxy-4-methylthio phenyl 4-phenylmethyl-1-piperidinyl, 2-methoxy-4-methylthiophenyl 4-benzylpiperidyl ketone, 1-2-methoxy-4-methylthio benzoyl-4-benzylpiperidine, 4-benzylpiperidino 2-methoxy-4-methylsulfanyl phenyl methanone, 1-2-methoxy-4-methylthio benzoyl-4-phenylmethyl piperidine
(4-benzylpiperidin-1-yl)-(2-methoxy-4-methylsulfanylphenyl)methanone
C21H25NO2S
OMGLGPKQUFSRNN-UHFFFAOYSA-N
1126109
COC1=C(C=CC(=C1)SC)C(=O)N2CCC(CC2)CC3=CC=CC=C3

Specifications

10mg
349087-34-9
unii-tbu7utv30q, tbu7utv30q, 4-benzylpiperidin-1-yl 2-methoxy-4-methylsulfanyl phenyl methanone, 4-benzyl-1-2-methoxy-4-methylsulfanyl benzoyl piperidine, piperidine, 1-2-methoxy-4-methylthio benzoyl-4-phenylmethyl, methanone, 2-methoxy-4-methylthio phenyl 4-phenylmethyl-1-piperidinyl, 2-methoxy-4-methylthiophenyl 4-benzylpiperidyl ketone, 1-2-methoxy-4-methylthio benzoyl-4-benzylpiperidine, 4-benzylpiperidino 2-methoxy-4-methylsulfanyl phenyl methanone, 1-2-methoxy-4-methylthio benzoyl-4-phenylmethyl piperidine
COC1=C(C=CC(=C1)SC)C(=O)N2CCC(CC2)CC3=CC=CC=C3
355.496
355.49
JX 401
C21H25NO2S
OMGLGPKQUFSRNN-UHFFFAOYSA-N
(4-benzylpiperidin-1-yl)-(2-methoxy-4-methylsulfanylphenyl)methanone
1126109
>98%
Videos
Documents