missing translation for 'onlineSavingsMsg'
Learn More

JW 480, Tocris Bioscience™

Potent and selective inhibitor of serine hydrolase KIAA1363 (AADACL1)

Brand:  RD Systems (Bio-Techne) 4327/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13444564

  • £129.36 / 10mg

Estimated Shipment date
from Supplier 28-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective inhibitor of the serine hydrolase enzyme KIAA1363 (AADACL1) (IC50 = 20 nM in mouse brain). Impairs migration, invasion, survival and tumor growth of prostate cancer cell lines in vivo. Reduces monoalkylglycerol ether (MAGE) levels.
Specifications

Chemical Identifiers

1354359-53-7
333.431
2-isopropylphenyl 2-naphthalen-2-yl ethyl carbamate, 2-isopropylphenyl n-2-naphthalen-2-yl ethyl carbamate
(2-propan-2-ylphenyl) N-(2-naphthalen-2-ylethyl)carbamate
C22H23NO2
PCNJGBMWAZRVEA-UHFFFAOYSA-N
57330099
CC(C)C1=CC=CC=C1OC(=O)NCCC2=CC3=CC=CC=C3C=C2

Specifications

10mg
1354359-53-7
2-isopropylphenyl 2-naphthalen-2-yl ethyl carbamate, 2-isopropylphenyl n-2-naphthalen-2-yl ethyl carbamate
CC(C)C1=CC=CC=C1OC(=O)NCCC2=CC3=CC=CC=C3C=C2
333.431
333.42
JW 480
C22H23NO2
PCNJGBMWAZRVEA-UHFFFAOYSA-N
(2-propan-2-ylphenyl) N-(2-naphthalen-2-ylethyl)carbamate
57330099
>98%
Videos
Documents