missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Plunger Dispensing Can

Justrite Plunger Dispensing Can provides a quick and safe means to moisten cloths with flammable cleaning fluids. Made of chemically-resistant Ryton™ and brass, the pump base is designed to leave less liquid in the bottom before having to refil.

Brand:  Justrite™ 10308Z

Code : 03

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 12981395

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Steel construction with durable, lead-free chemical-resistant powder paint
  • “Smart pump” mechanism helps streamline swabbing operations
  • Place cloth on tray and push down to wet cloth
  • Excess liquid safely drains back into reservoir
  • Plunger is made of chemically resistant Ryton™ and brass and is designed to leave less liquid in the bottom before refilling
  • Perforated pan screen serves as flame arrester
  • Easy to dismantle and reassemble for routine cleaning
  • Available in high-density polyethylene or rugged 24-gauge steel construction

Specifications

Specifications

FM, TUV
For dispensing flammable liquids
Brass and Ryton Plunger Assembly
4L
Steel
184mm
267mm
Plunger Can
Red
Videos