missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Sure-Grip™ Piggyback Flammable Safety Cabinet
Designed to mount on top of 30- and 45 gallon Sure-Grip™ EX cabinets, these Piggyback safety cabinets can be used independently or stacked as benchtop units for added storage.
£778.26
Specifications
Depth (English) Interior | 14.56 in. |
---|---|
Depth (Metric) Exterior | 45.7 cm |
Depth (Metric) Interior | 37 cm |
Color | Yellow |
Depth (English) Exterior | 18 in. |
Product Code | Brand | Certifications/Compliance | Dimensions (D x W x H) Interior | Height (Metric) Exterior | Weight (Metric) | Door Style | Price | Quantity & Availability | |
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Certifications/Compliance | Dimensions (D x W x H) Interior | Height (Metric) Exterior | Weight (Metric) | Door Style | Price | Quantity & Availability | |
11965366
|
Justrite™
8913201 |
FM, NFPA, OSHA, U (NFPA) | 14.56 x 39.5 x 13 in. (37 x 100.3 x 33 cm) | 45.7 cm | 59.4 kg | Self-close |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||
Description
Piggyback Safety Cabinet Features
- Stack atop standard 30 gal. (114L) safety cabinets
- Can also be wall mounted or used on countertop
- 18-gauge steel with 1.5 in. (3.8cm) insulating air space
- Available with manual or self-closing doors with flush-mounted Sure-Grip™ handles with exclusive gripper pad for secure hold and security
- Double-wall construction provides 1 1/2 in. of insulating air space to help prevent the spread of flames
- High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
- Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- SpillSlope safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Backed by an exclusive 10-year warranty
- Meet OSHA and NFPA requirements
Certifications
FM approved. Self-closing models also UL listed.
Compliance
Comply with Uniform Fire Code.
Specifications
14.56 in. | |
37 cm | |
18 in. | |
Piggyback storage cabinet | |
Flammables | |
2 | |
39.5 in. | |
100.3 cm |
45.7 cm | |
Yellow | |
Steel (CR) | |
43 in. | |
Flush mounted Sure-Grip™ handles with cylinder lock and key set or an optional padlock, Haz-Alert™ reflective trilingual warning labels, dual vents with flame arresters, four adjustable self leveling feet, grounding con | |
Epoxy, Polyester | |
109 cm |