missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ Piggyback Flammable Safety Cabinet

Designed to mount on top of 30- and 45 gallon Sure-Grip™ EX cabinets, these Piggyback safety cabinets can be used independently or stacked as benchtop units for added storage.

Brand:  Justrite™ 8913001

Code : 03

Additional Details : Weight : 58.51341kg

 View more versions of this product

Product Code. 11955356

  • £763.49 / Each


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Piggyback Safety Cabinet Features

  • Stack atop standard 30 gal. (114L) safety cabinets
  • Can also be wall mounted or used on countertop
  • 18-gauge steel with 1.5 in. (3.8cm) insulating air space
  • Available with manual or self-closing doors with flush-mounted Sure-Grip™ handles with exclusive gripper pad for secure hold and security
Features of Justrite Flammables Cabinets
  • Double-wall construction provides 1 1/2 in. of insulating air space to help prevent the spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • SpillSlope safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Backed by an exclusive 10-year warranty
  • Meet OSHA and NFPA requirements

Specifications

Specifications

12 gal.
45.7cm
14.56 x 39.5 x 13 in. (37 x 100.3 x 33cm)
Flammables
33cm
2
Piggyback storage cabinet
109cm
Yellow
FM, NFPA, OSHA
37cm
Manual-close
45.7cm
Flush mounted Sure-Grip™ handles with cylinder lock and key set or an optional padlock, Haz-Alert™ reflective trilingual warning labels, dual vents with flame arresters, four adjustable self leveling feet, grounding con
Epoxy, Polyester
59kg
100.3cm
Steel (CR)
Videos
View accessory products