missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Sure-Grip™ EX Compac Flammable Safety Cabinet

Justrite Compac Safety Cabinets fit nearly anywhere and provide protection where space is at a premium. The cabinets protect workers, reduce fire risks, and allow storage of flammable fuels and chemicals in code-compliant safety cabinets.

Brand:  Justrite™ 8912201

Code : 03

Additional Details : Weight : 55.33826kg

 View more versions of this product

Product Code. 11915356

  • £741.34 / Each


Estimated Shipment
02-08-2018
Add to basket

Includes: One adjustable shelf.

Description

Description

  • One-door cabinet stores up to 12 gal. (45.4L) of flammables
  • Double-wall construction provides 11/2 in. of insulating air space to help prevent the spread of flames Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • High-visibility Haz-Alert™ reflective warning labels provide added safety during power outages or fires
  • Self-latching doors facilitate fire protection of the cabinet
  • Self latch engages critical three-point latching system for protection under fire conditions
  • Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents

  • One adjustable SpillSlope™ safety shelf directs spills to back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Finished inside and out with a durable lead-free hybrid epoxy and polyester powder paint for chemical resistance
  • Fit neatly under counter or stand alone
  • Lettering on door reads: “FLAMMABLE—KEEP FIRE AWAY”
  • Backed by an exclusive 10-year warranty
  • Meet OSHA and NFPA requirements

19-121-852: Manual-close door
19-121-853; Self-closing door

Certifications

FM approved. Self-closing model UL listed.

Compliance

Meet NFPA and OSHA requirements. Self-closing model complies with Uniform Fire Code.

Specifications

Specifications

12 gal.
45.7cm
14.56 x 20 x 30.5 in. (37 x 50.8 x 77.5cm)
Flammables
77.5cm
1
Epoxy, Polyester
69kg
50.8cm
Steel (CR)
FM, NFPA, OSHA
37cm
Self-closing
88.9cm
One adjustable shelf, U-Loc™ handle with two keys and accept padlock, Haz-Alert™ reflective exterior labeling and Galvanized Steel SpillSlope™ shelves, leakproof 2 in. bottom sump, dual vents with flam
1
Storage Cabinet
59.05cm
Yellow
Videos