missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Elite Smokers Cease-Fire Cigarette Butt Receptacles Accessories

Brand:  Justrite™ 268506

Code : 03

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12990225

  • £11.33 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Anchoring Cable: 91.4cm (36 in.) wire rope cable provides added security or greater stability in severe weather
  • Brass Padlock: 3/16 in. dia. shackle
  • Bucket Liners: special, burn-resistant laminated aluminum/poly construction; include twist-ties; fit any bucket 29.8cm (11.75 in.) wide x 25.4cm (10 in.) high maximum size

Specifications

Specifications

Justrite™ elite smokers cease-fire cigarette butt receptacles
Brass
Padlock
Each
Videos