missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ EcoPolyBlend™ Drum Management System
Features a space-saving stackable design
Brand: Justrite™ 28667
Code : 03
Additional Details : Weight : 62.70000kg
Description
100% molded polyethylene construction provides maximum chemical resistance, preventing corrosion, deterioration and leaks
- Stores up to four 110 and/or 200L (30 and/or 55 gal.) drums
- Stores drums of Class 1B or 1C flammables and combustibles or nonhazardous liquids
- Consists of three parts: a base module, stack module and dispensing shelf
- Molded-in drum lock-channels hold drums in place
Base Module
- Holds two drums with a 1388kg (3060 lb.) total load capacity
- Easily relocates with an empty forklift
- 250L (66 gal.) open-view containment sump makes it easy to spot any spills for a quick cleanup
- Holds two additional drums to double the system capacity for a load of 680kg (1500 lb.)
- When storing both 110 and 200L (30 and 55-gal.) drums, the smaller drums should be placed on the top of the stack module
- Mounts securely to the stack module for convenient dispensing from the upper level
- Has a load capacity of 27 kg (60 lb.)
Compliance
EPA, NFPA, OSHA, SPCC
Specifications
SPCC, EPA, NFPA, OSHA | |
66 gal. | |
Polyethylene | |
Efficient storage, positive spill control, liquid dispensing | |
Dispensing well | |
1388 kg | |
124.46 cm | |
66 cm | |
149.8 cm | |
2.5 cm | |
0.45 kg |
Black | |
249.8 L | |
Drum Management Base Module | |
2 Drums | |
3060 lb. | |
49 in. | |
26 in. | |
59 in. | |
1 in. | |
1 lb. |