Learn More
3M™ Jupiter™ Ready-To-Use Starter Kits
Ready-to-use starter kits feature everything needed for filtration in the laboratory. Complete with electronic audible and visual alarms in the event of low battery and/or low air flow.
Brand: 3M™ 7100079779
Code : NEW
Additional Details : Weight : 3.71700kg
Description
- High comfort decontamination belt
- Air Flow Indicator and Calibration tube
- Pair of Organic Vapour and Particulate (A2P) Filters and Prefilters
- Length-adjusting breathing tube
- 8-hour rechargeable battery (NiMH) and a battery charger
Specifications
8 hours | |
OV/HE Filter | |
Protection against gases, vapors, and particulates, or a combination of contaminants per filter used | |
JPRTU1 |
NiMH | |
PAPR Assembly, No Headtop | |
Powered turbo unit, decontamination belt, calibration tube and airflow indicator, 2 x A2P filters, pre-filters, breathing tube, rechargeable battery, battery charger | |
1 Ea. |