missing translation for 'onlineSavingsMsg'
Learn More

Julolidine hydrobromide, 97%, Thermo Scientific™

Catalog Number p-7052542
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
83646-41-7
C12H16BrN
254.17
MFCD00198009
KHWBRFVBPGPEOJ-UHFFFAOYSA-N
julolidine hydrobromide, 2,3,6,7-tetrahedron-1h,5h-pyrido 3,2,1-ij quinoline hydrobromide, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine hydrobromide, labotest-bb lt00451605, 2,3,6,7-tetrahydro-1h-5h-benzo i,j quinolizine, hbr, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline hydrobromide
12255670
1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene hydrobromide
Br.C1CN2CCCC3=C2C(C1)=CC=C3
This item is not returnable. View return policy
83646-41-7
C12H16BrN
254.17
MFCD00198009
KHWBRFVBPGPEOJ-UHFFFAOYSA-N
julolidine hydrobromide, 2,3,6,7-tetrahedron-1h,5h-pyrido 3,2,1-ij quinoline hydrobromide, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine hydrobromide, labotest-bb lt00451605, 2,3,6,7-tetrahydro-1h-5h-benzo i,j quinolizine, hbr, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline hydrobromide
12255670
1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene hydrobromide
Br.C1CN2CCCC3=C2C(C1)=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 239°C to 242°C
Color Pink
Quantity 5 g
UN Number UN2923
Sensitivity Hygroscopic
Formula Weight 254.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Julolidine hydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'