missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Julolidine hydrobromide, 97%

Brand:  Alfa Aesar™ H36298.14

Code : NEW

Additional Details : CAS Number : 83646-41-7 Weight : 0.02500kg Transport : UN number : 2923 Chem class : 1 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 15477637

  • £93.40 / 25g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Julolidine hydrobromide
254.17
UN2923
83646-41-7
MFCD00198009
2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine hydrobromide

Specifications

Julolidine hydrobromide
Pale pink
25g
Solid
254.17
UN2923
Hygroscopic
239° to 242°C
83646-41-7
97%
MFCD00198009
2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine hydrobromide
Videos
SDS