missing translation for 'onlineSavingsMsg'
Learn More

JTC 801, Tocris Bioscience™

Selective NOP antagonist

Brand:  Tocris (Bio-Techne) 248150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461304

  • £583.59 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity, selective NOP receptor antagonist (Ki = 8.2 nM). Displays approximately 12.5-, 129- and 1055-fold selectivity over human μ-, κ- and δ-opioid receptors respectively. Exhibits anti-nociceptive effects in acute pain models in vivo. Orally active.
Specifications

Chemical Identifiers

244218-51-7
447.963
unii-7i21wlz2fp, 7i21wlz2fp, n-4-amino-2-methyl-6-quinolinyl-2-4-ethylphenoxy methyl benzamide hydrochloride, n-4-amino-2-methylquinolin-6-yl-2-4-ethylphenoxy methyl benzamide hydrochloride, n-4-amino-2-methylquinolin-6-yl-2-4-ethylphenoxymethyl benzamide hydrochloride, d0f1ef, jtc-801 hcl, auryntricarboxylic acid ata, benzamide, n-4-amino-2-methyl-6-quinolinyl-2-4-ethylphenoxy methyl-, monohydrochloride
N-(4-amino-2-methylquinolin-6-yl)-2-[(4-ethylphenoxy)methyl]benzamide;hydrochloride
C26H26ClN3O2
NQLIYKXNAXKMBL-UHFFFAOYSA-N
5311339
CCC1=CC=C(C=C1)OCC2=CC=CC=C2C(=O)NC3=CC4=C(C=C3)N=C(C=C4N)C.Cl

Specifications

50mg
244218-51-7
unii-7i21wlz2fp, 7i21wlz2fp, n-4-amino-2-methyl-6-quinolinyl-2-4-ethylphenoxy methyl benzamide hydrochloride, n-4-amino-2-methylquinolin-6-yl-2-4-ethylphenoxy methyl benzamide hydrochloride, n-4-amino-2-methylquinolin-6-yl-2-4-ethylphenoxymethyl benzamide hydrochloride, d0f1ef, jtc-801 hcl, auryntricarboxylic acid ata, benzamide, n-4-amino-2-methyl-6-quinolinyl-2-4-ethylphenoxy methyl-, monohydrochloride
CCC1=CC=C(C=C1)OCC2=CC=CC=C2C(=O)NC3=CC4=C(C=C3)N=C(C=C4N)C.Cl
447.963
447.96
JTC 801
C26H26ClN3O2
NQLIYKXNAXKMBL-UHFFFAOYSA-N
N-(4-amino-2-methylquinolin-6-yl)-2-[(4-ethylphenoxy)methyl]benzamide;hydrochloride
5311339
>98%
Videos
Documents