missing translation for 'onlineSavingsMsg'
Learn More

JNJ 303, Tocris Bioscience™

Potent and selective IKs blocker

Brand:  Tocris (Bio-Techne) 389950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13443784

  • £618.21 / 50mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent IKs blocker (IC50 = 64 nM). Displays no effects on other cardiac channels; IC50 values are 3.3, >10, 11.1 and 12.6 μM for INa, ICa, Ito and IKr currents respectively. Prolongs QT and causes unprovoked torsades de pointes (TdP).
Specifications

Chemical Identifiers

878489-28-2
440.983
2-(4-Chlorophenoxy)-2-methyl-N-[5-[(methylsulfonyl)amino]tricyclo[3.3.1.13, 7]dec-2-yl]-propanamide
2-(4-chlorophenoxy)-N-[(1R,3S)-5-(methanesulfonamido)-2-adamantyl]-2-methylpropanamide
C21H29ClN2O4S
OSGIRCJRKSAODN-DJASPMHUSA-N
11575823
CC(C)(C(=O)NC1C2CC3CC1CC(C3)(C2)NS(=O)(=O)C)OC4=CC=C(C=C4)Cl

Specifications

JNJ 303
878489-28-2
2-(4-Chlorophenoxy)-2-methyl-N-[5-[(methylsulfonyl)amino]tricyclo[3.3.1.13, 7]dec-2-yl]-propanamide
CC(C)(C(=O)NC1C2CC3CC1CC(C3)(C2)NS(=O)(=O)C)OC4=CC=C(C=C4)Cl
440.983
440.98
50mg
C21H29ClN2O4S
OSGIRCJRKSAODN-DJASPMHUSA-N
2-(4-chlorophenoxy)-N-[(1R,3S)-5-(methanesulfonamido)-2-adamantyl]-2-methylpropanamide
11575823
>98%
Videos