missing translation for 'onlineSavingsMsg'
Learn More

JNJ 10191584 maleate, Tocris Bioscience™

Selective H4 receptor antagonist; orally active

Brand:  Tocris (Bio-Techne) 244110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461214

  • £133.65 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Highly selective histamine H4 receptor silent antagonist; binds with high affinity to the human H4 receptor (Ki = 26 nM). > 540-fold selective over the H3 receptor (Ki = 14.1 μM). Inhibits mast cell and eosinophil chemotaxis in vitro with IC50 values of 138 and 530 nM respectively. Orally active in vivo.
Specifications

Chemical Identifiers

869497-75-6
396.828
jnj 10191584 maleate
(6-chloro-1H-benzimidazol-2-yl)-(4-methylpiperazin-1-yl)methanone;formic acid;prop-2-enoic acid
C17H21ClN4O5
BODOIXYJOORQBQ-UHFFFAOYSA-N
77519402
CN1CCN(CC1)C(=O)C2=NC3=C(N2)C=C(C=C3)Cl.C=CC(=O)O.C(=O)O

Specifications

JNJ 10191584 maleate
869497-75-6
jnj 10191584 maleate
CN1CCN(CC1)C(=O)C2=NC3=C(N2)C=C(C=C3)Cl.C=CC(=O)O.C(=O)O
396.828
394.81
10mg
C17H21ClN4O5
BODOIXYJOORQBQ-UHFFFAOYSA-N
(6-chloro-1H-benzimidazol-2-yl)-(4-methylpiperazin-1-yl)methanone;formic acid;prop-2-enoic acid
77519402
>99%
Videos