missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse JMJD6/PSR Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | JMJD6/PSR |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
JMJD6/PSR Polyclonal specifically detects JMJD6/PSR in Mouse samples. It is validated for Western Blot.Specifications
JMJD6/PSR | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
bifunctional arginine demethylase and lysyl-hydroxylase JMJD6, EC 1.14.11, EC 1.14.11.-, Histone arginine demethylase JMJD6, JmjC domain-containing protein 6, jumonji domain containing 6, Jumonji domain-containing protein 6, KIAA0585PSR, Lysyl-hydroxylase JMJD6, Peptide-lysine 5-dioxygenase JMJD6, Phosphatidylserine receptorPTDSR, Protein PTDSR, PSR, PTDSR, PTDSR1 | |
JMJD6 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9ERI5 | |
23210 | |
Mouse myeloma cell line NS0-derived recombinant mouse JMJD6/PSR Ala289-Arg403 Accession # Q9ERI5 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse JMJD6/PSR in direct ELISAs and Western blots. |
For Research Use Only