missing translation for 'onlineSavingsMsg'
Learn More

JK 184, Tocris Bioscience™

Alcohol dehydrogenase 7 inhibitor; inhibits downstream Hh signaling pathway

Brand:  Tocris (Bio-Techne) 334150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13452844

  • £645.33 / 50mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent downstream hedgehog (Hh) signaling inhibitor that prevents Gli-dependent transcriptional activity (IC50 = 30 nM). Exhibits antiproliferative activity in a range of cancer cell lines (GI50 = 3 - 21 nM) and in human xenografts in vivo. Inhibits alcohol dehydrogenase 7 (Adh7) (IC50 = 210 nM) and acts as a microtubule depolymerizing agent in vitro.
Specifications

Chemical Identifiers

315703-52-7
350.44
n-4-ethoxyphenyl-4-2-methylimidazo 1,2-a pyridin-3-yl thiazol-2-amine, hh signaling antagonist vii, jk184, n-4-ethoxyphenyl-4-2-methylimidazo 1,2-a pyridin-3-yl-2-thiazolamine, n-4-ethoxyphenyl-4-2-methylimidazo 1,2-a pyridin-3-yl-1,3-thiazol-2-amine, shh antagonist, hh signaling antagonist vii, 3-2-4-ethoxyphenyl amino thiazole-4-yl-2-methylimidazo 1,2-a pyridine, 2-thiazolamine, n-4-ethoxyphenyl-4-2-methylimidazo 1,2-a pyridin-3-yl, n-4-ethoxyphenyl-4-2-methylimidazo-1,2-a pyridin-3-yl thiazol-2-amine
N-(4-ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)-1,3-thiazol-2-amine
C19H18N4OS
ROYXIPOUVGDTAO-UHFFFAOYSA-N
1069686
CCOC1=CC=C(C=C1)NC2=NC(=CS2)C3=C(N=C4N3C=CC=C4)C

Specifications

50mg
315703-52-7
n-4-ethoxyphenyl-4-2-methylimidazo 1,2-a pyridin-3-yl thiazol-2-amine, hh signaling antagonist vii, jk184, n-4-ethoxyphenyl-4-2-methylimidazo 1,2-a pyridin-3-yl-2-thiazolamine, n-4-ethoxyphenyl-4-2-methylimidazo 1,2-a pyridin-3-yl-1,3-thiazol-2-amine, shh antagonist, hh signaling antagonist vii, 3-2-4-ethoxyphenyl amino thiazole-4-yl-2-methylimidazo 1,2-a pyridine, 2-thiazolamine, n-4-ethoxyphenyl-4-2-methylimidazo 1,2-a pyridin-3-yl, n-4-ethoxyphenyl-4-2-methylimidazo-1,2-a pyridin-3-yl thiazol-2-amine
CCOC1=CC=C(C=C1)NC2=NC(=CS2)C3=C(N=C4N3C=CC=C4)C
350.44
350.44
JK 184
C19H18N4OS
ROYXIPOUVGDTAO-UHFFFAOYSA-N
N-(4-ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)-1,3-thiazol-2-amine
1069686
>99%
Videos
Documents