Learn More
JIP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557057
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FSLSTFHSLS PPGCRPPQDI SLEEFDDEDL SEITDDCGLG LSYDSDHCEK DSLSLGRSEQ PHPICSFQDD FQEFEM Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene is closely related to MAPK8IP1/IB1/JIP-1, a scaffold protein that is involved in the c-Jun amino-terminal kinase signaling pathway. This protein is expressed in brain and pancreatic cells. It has been shown to interact with, and regulate the activity of MAPK8/JNK1, and MAP2K7/MKK7 kinases. This protein thus is thought to function as a regulator of signal transduction by protein kinase cascade in brain and pancreatic beta-cells. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene.Specifications
JIP2 | |
Polyclonal | |
Unconjugated | |
MAPK8IP2 | |
3230402N03Rik; AI847694; AW049958; C-Jun-amino-terminal kinase-interacting protein 2; homologous to mouse JIP-1; IB 2; Ib2; IB-2; Islet brain 2; islet-brain 2; islet-brain-2; JIP 2; JIP2; JIP-2; JNK MAP kinase scaffold protein 2; JNK MAP kinase scaffold protein JIP2; JNK-interacting protein 2; Mapk8ip2; mitogen activated protein kinase 8 interacting protein 2; mitogen-activated protein kinase 8 interacting protein 2; mitogen-activated protein kinase 8-interacting protein 2; PRKM8IPL; R75148; si:dkey-180h12.2; si:dkey-181m21.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23542 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13387 | |
MAPK8IP2 | |
Recombinant protein corresponding to Human JIP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only