missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Jenway™ Interference Filters
Better performance on your spectrophotometers
£443.00 - £1009.76
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
11404879
|
Cole-Parmer™
99968-79 |
360 nm |
£1009.76
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11464889
|
Cole-Parmer™
15986-41 |
480 nm |
£496.93
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
11414899
|
Cole-Parmer™
99968-71 |
520 nm |
£496.93
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11404889
|
Cole-Parmer™
10089-83 |
430 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11494879
|
Cole-Parmer™
10089-82 |
420 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11464899
|
Cole-Parmer™
99968-72 |
550 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11404919
|
Cole-Parmer™
16181-19 |
660 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11424919
|
Cole-Parmer™
10090-04 |
680 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11474879
|
Cole-Parmer™
10089-80 |
410 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Choose accessories for your application
- Easy to install
Specifications
Interference Filter |