missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ JC-1 mitochondrial membrane potential dependent fluorescent dye
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Purity:>98% (HPLC); NMR (conforms)
- CAS No.: 3520-43-2
- Molecular Formula: C25H27Cl4IN4
- Molecular Weight:652.2
- Source:Synthetic
- Appearance:Red powder
- PubChem ID: 5492929
Specifications
Specifications
| Format | Red powder |
| Quantity | 1 mg |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction