missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Jars

Brand:  Duran™ 242085701

Code : A0

Additional Details : Weight : 4.13000kg

 View more versions of this product

Product Code. 12625706

  • £681.60 / Pack of 10


Estimated Shipment
28-03-2018
Add to basket

Specifications

Specifications

1000mL
10Pack
10mL
150mm
Lid
10Pack
Jar
Videos