Learn More
Tocris Bioscience™ Janelia Fluor™ 635, Maleimide
Description
Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) including dSTORM (in both live and fixed cells). Cell permeable.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 645 nm and 664 nm, respectively.
Please see the product protocol for further information and a guide to protein/antibody labeling.
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company R&DSystems.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 4-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)carbamoyl)-2-(3-(3-fluoroazetidin-1-ium-1-ylidene)-7-(3-fluoroazetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)benzoate |
| Quantity | 1 mg |
| Target | Fluorescent Dyes |
| Molecular Formula | C35H32F2N4O5Si |
| Purity | 0.95 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.