Learn More
JAML Monoclonal Antibody (eBio4E10 (4E10)), eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 14-5666-81
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The eBio4E10 monoclonal antibody reacts with mouse JAML (junctional adhesion molecule-like protein), also known as AMICA. JAML is a glycosylated membrane protein possessing two Ig-like domains in its extracellular domain. JAML is expressed on polymorphonuclear leukocytes (PMN), and is involved in the transmigration of such cells through epithelial tight junctions via interactions between JAML and epithelial coxsackie and adenovirus receptor (CAR). JAML is also expressed on lymphocyte subsets, such as gammadelta T cell lines, and can provide co-stimulatory signals for cell activation. Applications Reported: This eBio4E10 antibody has been reported for use in flow cytometric analysis and immunoprecipitation. Applications Tested: This eBio4E10 (4E10) antibody has been tested by flow cytometric analysis of mouse lysed whole blood. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
JAML (junctional adhesion molecule-like protein), also known as AMICA. JAML is a glycosylated membrane protein possessing two Ig-like domains in its extracellular domain. JAML is expressed on polymorphonuclear leukocytes (PMN), and is involved in the transmigration of such cells through epithelial tight junctions via interactions between JAML and epithelial coxsackie and adenovirus receptor (CAR). JAML is also expressed on lymphocyte subsets, such as gamma-delta T cell lines, and can provide co-stimulatory signals for cell activation.Specifications
JAML | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q80UL9 | |
Jaml | |
Affinity chromatography | |
RUO | |
270152 | |
4° C | |
Liquid |
Flow Cytometry, Immunoprecipitation | |
eBio4E10 (4E10) | |
Unconjugated | |
Jaml | |
AMICA1, CREA7-1, JAML, adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
Armenian Hamster | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
For Research Use Only.