missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM-A Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
POA
Specifications
Antigen | JAM-A |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse JAM-A Lys27-Ala242 Accession # O88792 |
Gene ID (Entrez) | 50848 |
Gene Accession No. | O88792 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
JAM-A Polyclonal specifically detects JAM-A in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).Specifications
JAM-A | |
50848 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse JAM-A in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human JAM-A, recombinant mouse (rm) JAM-B, and rmJAM-C is observed. | |
Unconjugated | |
Western Blot, Immunohistochemistry, ELISA | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant mouse JAM-A Lys27-Ala242 Accession # O88792 | |
O88792 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only