Learn More
Kimberly Clark™ Jackson™ Safety Respirators
Brand:  Kimberly Clark™ 62970
UNSPSC : 46182007
Code : 02
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12932415
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications Description & Specifications
Specifications
10 pack
Valved (carbon) and folded
Resisting nuisance odors, filtering fine dust, and water and oil-based mist; providing medium levels of protection (factor 12) against pollution, easier breathing when working in moderately contaminated or dusty areas, shaping easily to suit your face; keeping your face cooler and drier.
Valved Carbon Respirator
Yellow
EN149:2001+A1:2009