Learn More
Kimberly Clark™ Jackson™ Safety Respirators
Brand:  Kimberly Clark™ 62980
UNSPSC : 46182007
Code : 02
Additional Details : Weight : 1.00000kg
Product Code. 12942415
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications Description & Specifications
Specifications
10 pack
Valved and folded
Yes
R30 FFP3 RD
Folded Respirator
Orange
Filtering metal fumes, fine dust, water and oil-based mist, caustic substances and solvents, providing high levels of protection (factor 50) against pollution, easier breathing when working in heavily contaminated or dusty areas; protection for longer periods without clogging; keeping your face cooler and drier
EN149:2001+A1:2009