Learn More
Kimberly Clark™ Jackson™ Safety Respirators
Brand:  Kimberly Clark™ 64570
UNSPSC : 46182007
Code : S7
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12472320
  • £386.00 / Pack of 80
Estimated Shipment
19-06-2017
Add to basket
Specifications Description & Specifications
Specifications
R30 FFP3 NRD
EN149:2001+A1:2009
Red
10 pack
Molded and dual valved
Dual Valved Respirator
Yes
Filtering fine dust, water- and oil-based mist, and metal fumes; providing high levels of protection (factor 50) against pollution; easier breathing when working in heavily contaminated or dusty areas; keeping your face cooler and drier; shaping easily to suit your face.