Learn More
Kimberly-Clark™ Jackson™ Safety Respirators
Brand: Kimberly-Clark™ 62940
Code : 02
Additional Details : Weight : 1.00000kg
Specifications
10 pack | |
Unvalved and folded | |
Filtering fine dust, and water and oil based mist when working in moderately contaminated or dusty areas; providing medium levels of protection (factor 12) against pollution; protecting for longer periods without clogging; shaping easily to suit your face; wearing with Jackson safety Eye Wear without making lenses misty. | |
EN149:2001+A1:2009 |
Folded Respirator | |
Yellow | |
R20 FFP2 RD |