Learn More
Kimberly Clark™ Jackson™ Safety Respirators
Brand:  Kimberly Clark™ 62940
Code : NEW
Additional Details : Weight : 0.80000kg
Product Code. 11803540
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
10 pack
Unvalved and folded
Filtering fine dust, and water and oil based mist when working in moderately contaminated or dusty areas; providing medium levels of protection (factor 12) against pollution; protecting for longer periods without clogging; shaping easily to suit your face; wearing with Jackson safety Eye Wear without making lenses misty.
EN149:2001+A1:2009
Folded Respirator
Yellow
R20 FFP2 RD
Videos