Learn More
Kimberly Clark™ Jackson™ Safety Respirators
Brand:  Kimberly Clark™ 62920
UNSPSC : 46182007
Code : 02
Additional Details : Weight : 0.01000kg
Product Code. 12515478
  • £274.00 / Pack of 120
Estimated Shipment
03-08-2017
Add to basket
Specifications Description & Specifications
Specifications
10 pack
Unvalved and folded
Filtering fine dust, and water- and oil-based mist when working in moderately contaminated or dusty areas; providing protection (factor 4) against pollution;
EN149:2001+A1:2009
Folded Respirator
Blue
R10 FFP1 NR