Kimberly Clark™ Jackson™ Safety Respirators
Overview
Brand: Kimberly Clark™
Manufacturer Part Number: 62920
UNSPSC: 41120000
Code: NEW
Additional Details:
Additional Details: Weight: 0.80000kg
Product Code. 11813540
 N/A
N/A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description and Specification
Specification
Construction Unvalved and folded
Model R10 FFP1 NR
Certifications EN149:2001+A1:2009
Product Type Folded Respirator
Color Blue
Quantity 10 pack
For Use With (Application) Filtering fine dust, and water- and oil-based mist when working in moderately contaminated or dusty areas; providing protection (factor 4) against pollution;