missing translation for 'onlineSavingsMsg'
Learn More

(±)-J 113397, Tocris Bioscience™

Potent and selective NOP antagonist

Brand:  Tocris (Bio-Techne) 259810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441544

  • £244.21 / 10mg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective NOP receptor antagonist (IC50 values are 2.3, 1400, 2200 and > 10000 nM for NOP, κ, μ and δ-opioid receptors respectively). Inhibits nociceptin/orphanin FQ-induced hyperalgesia in the mouse tail-flick test.
Specifications

Chemical Identifiers

217461-40-0
399.579
(±)-1-[(3R™, 4R™)-1-(Cyclooctylmethyl)-3-(hydroxymethyl)-4-piperidinyl]-3-ethyl-1, 3-dihydro-2H-benzimidazol-2-one
1-[1-(cyclooctylmethyl)-3-(hydroxymethyl)piperidin-4-yl]-3-ethylbenzimidazol-2-one
C24H37N3O2
MBGVUMXBUGIIBQ-UHFFFAOYSA-N
21186500
CCN1C2=CC=CC=C2N(C1=O)C3CCN(CC3CO)CC4CCCCCCC4

Specifications

(±)-J 113397
217461-40-0
(±)-1-[(3R™, 4R™)-1-(Cyclooctylmethyl)-3-(hydroxymethyl)-4-piperidinyl]-3-ethyl-1, 3-dihydro-2H-benzimidazol-2-one
CCN1C2=CC=CC=C2N(C1=O)C3CCN(CC3CO)CC4CCCCCCC4
399.579
399.57
10mg
C24H37N3O2
MBGVUMXBUGIIBQ-UHFFFAOYSA-N
1-[1-(cyclooctylmethyl)-3-(hydroxymethyl)piperidin-4-yl]-3-ethylbenzimidazol-2-one
21186500
>98%
Videos