missing translation for 'onlineSavingsMsg'
Learn More

IWP 2, Tocris Bioscience™

PORCN inhibitor; inhibits Wnt processing and secretion

Brand:  Tocris (Bio-Techne) 353350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13443154

  • £723.24 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of Wnt processing and secretion. Inactivates PORCN, a membrane-bound O-acyltransferase (MBOAT), and selectively inhibits palmitoylation of Wnt. Blocks Wnt-dependent phosphorylation of Lrp6 receptor and Dvl2, and β-catenin accumulation. Suppresses self-renewal in R1 embryonic stem cells.
Specifications

Chemical Identifiers

686770-61-6
466.592
iwp-2, iwp 2, n-6-methyl-2-benzothiazolyl-2-3,4,6,7-tetrahydro-4-oxo-3-phenylthieno 3,2-d pyrimidin-2-yl thio-acetamide, n-6-methylbenzo d thiazol-2-yl-2-4-oxo-3-phenyl-3,4,6,7-tetrahydrothieno 3,2-d pyrimidin-2-yl thio acetamide, n-6-methyl-1,3-benzothiazol-2-yl-2-4-oxo-3-phenyl-6,7-dihydrothieno 3,2-d pyrimidin-2-yl sulfanyl acetamide, n-6-methylbenzo d thiazol-2-yl-2-4-oxo-3-phenyl-3,4,6,7-tetrahydrothieno 3,2-d pyrimidin-2-ylthio acetamide, n-6-methyl-1,3-benzothiazol-2-yl-2-4-oxo-3-phenyl-6h,7h-thieno 3,2-d pyrimidin-2-yl sulfanyl acetamide, iwp hplc, 9xk, n-6-methyl-2-benzothiazolyl-2-3,4,6,7-tetrahydro-4-oxo-3-phenylthieno 3,2-d pyrimidin-2-yl thio
N-(6-methyl-1,3-benzothiazol-2-yl)-2-[(4-oxo-3-phenyl-6,7-dihydrothieno[3,2-d]pyrimidin-2-yl)sulfanyl]acetamide
C22H18N4O2S3
WRKPZSMRWPJJDH-UHFFFAOYSA-N
2155128
CC1=CC2=C(C=C1)N=C(S2)NC(=O)CSC3=NC4=C(C(=O)N3C5=CC=CC=C5)SCC4

Specifications

50mg
686770-61-6
iwp-2, iwp 2, n-6-methyl-2-benzothiazolyl-2-3,4,6,7-tetrahydro-4-oxo-3-phenylthieno 3,2-d pyrimidin-2-yl thio-acetamide, n-6-methylbenzo d thiazol-2-yl-2-4-oxo-3-phenyl-3,4,6,7-tetrahydrothieno 3,2-d pyrimidin-2-yl thio acetamide, n-6-methyl-1,3-benzothiazol-2-yl-2-4-oxo-3-phenyl-6,7-dihydrothieno 3,2-d pyrimidin-2-yl sulfanyl acetamide, n-6-methylbenzo d thiazol-2-yl-2-4-oxo-3-phenyl-3,4,6,7-tetrahydrothieno 3,2-d pyrimidin-2-ylthio acetamide, n-6-methyl-1,3-benzothiazol-2-yl-2-4-oxo-3-phenyl-6h,7h-thieno 3,2-d pyrimidin-2-yl sulfanyl acetamide, iwp hplc, 9xk, n-6-methyl-2-benzothiazolyl-2-3,4,6,7-tetrahydro-4-oxo-3-phenylthieno 3,2-d pyrimidin-2-yl thio
CC1=CC2=C(C=C1)N=C(S2)NC(=O)CSC3=NC4=C(C(=O)N3C5=CC=CC=C5)SCC4
466.592
466.6
IWP 2
C22H18N4O2S3
WRKPZSMRWPJJDH-UHFFFAOYSA-N
N-(6-methyl-1,3-benzothiazol-2-yl)-2-[(4-oxo-3-phenyl-6,7-dihydrothieno[3,2-d]pyrimidin-2-yl)sulfanyl]acetamide
2155128
>98%
Videos
Documents